Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Grade GF/A Filter for Personal Air Samplers
These aerosol sampling and particulate monitoring filters provide high flow rates and minimal sample interference. Cytiva Whatman Grade GF/A card-mounted filters are used in static sample and personal air sampler applications.
$70.63
Specifications
Format Circular Disk
Material Borosilicate glass, card mount
View More Specs
Products 1
Catalog Number Mfr. No. Disc Diameter Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Disc Diameter Quantity Price Quantity & Availability  
09-920-095
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
1820900086
37mm
Pack of 80 for $70.63
Add to cart
Description
Description
  • Heat resistant (up to 500°C)
  • Made of high-quality borosilicate glass fibers without added binders
  • Low moisture affinity and high loading capacity
Specifications
Specifications
Circular Disk
Borosilicate glass, card mount