Learn More
Eisco™ Graduated Cylinder, Borosilicate, Class A, Round Base, 1.0ml grad
Borosilicate glass, Class - A, Round base with spout as per DIN EN ISO 4788 standard.
$5.85 - $6.55
Specifications
Material Borosilicate Glass
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
S15488 Products for Science Education
View Documents
Eisco™
CH0344L
100 mL
Each for $6.55
Add to cart
S15487 Products for Science Education
View Documents
Eisco™
CH0344K
50 mL
Each for $5.85
Add to cart
Specifications
Specifications
Borosilicate Glass