Learn More
Sklar InstrumentsSupplier Diversity Partner Graduated Measure Pitcher, Sklar, Plastic. This pitcher will hold up to 16 fluid ounces. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 101546
Graduated Measure Pitcher, Sklar, Plastic. Sklar's Graduated Measure Pitcher is a polyproylene container used to accurately measure fluids or measure and dilute chemicals. The pitcher features cubic centimeter and ounce markings. This pitcher will hold up to 16 fluid ounces.
Catalog No. 50-143-7058