Learn More
Sklar InstrumentsSupplier Diversity Partner Graduated Measure, Sklar, Stainless Steel. It has a 3-3/4 inch diameter base and a 4-5/8 in diameter opening. This pitcher holds 16 fluid oz. PK/12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 10155212
Graduated Measure, Sklar, Stainless Steel. Sklar's Stainless Steel Graduated Measure is a container used to accurately measure fluids or measure and dilute chemicals. The pitcher features cubic centimeter and ounce markings. It has a 3-3/4 inch diameter base and a 4-5/8 inch diameter opening. This pitcher will hold up to 16 fluid ounces. Pack of 12.
Catalog No. 50-143-7068