Learn More
Sklar InstrumentsSupplier Diversity Partner Graduated Measure, Sklar, Stainless Steel. It has a 3-3/4 inch diameter base and a 4-5/8 inch diameter opening. This pitcher holds 16 fluid oz. Pack of 6. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 1015526
Graduated Measure, Sklar, Stainless Steel. Sklar's Stainless Steel Graduated Measure is a container used to accurately measure fluids or measure and dilute chemicals. The pitcher features cubic centimeter and ounce markings. It has a 3-3/4 inch diameter base and a 4-5/8 inch diameter opening. This pitcher will hold up to 16 fluid ounces. Pack of 6.
Catalog No. 50-143-7069