Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graham-Field™ Lister Bandage Scissors
SDP

For medical and surgical procedures

$13.25 - $30.50

Specifications

Blade Style Straight
View More Specs

Products 3
Catalog Number Mfr. No. For Use With (Application) Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Length (English) Length (Metric) Price Quantity & Availability  
19-027486
SDP
View Documents
Graham-Field™
2606
Surgical, Precision 3.5 in. 9 cm
Each for $14.75
Add to Cart
 
19-027488
SDP
View Documents
Graham-Field™
2608
Bandage and Cast 5.5 in. 14 cm
Each for $13.25
Add to Cart
 
19-027489
SDP
View Documents
Graham-Field™
2609
Bandage and Cast 7.25 in. 18.5 cm
Each for $30.50
Add to Cart
 
Description

Description

  • Stainless steel
  • Available in a variety of sizes

Specifications

Specifications

Straight
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit