Graham-Field Stethoscopes

Manufacturer: GRAHAM FIELD INC 602NP

 View more versions of this product

Catalog No. 18-999-437D

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) To detect high, medium and low frequency sounds for infants, children and adults
    Includes Accessory pouch, interchangeable bells, adult and pediatric diaphragms, 2 pairs of eartips, 22 in. latex-free Y-tubing
    Color Neon Pink
    Product Type Sprague Rappaport Stethoscope

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit