Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Premium Grain Cowhide Leather Drivers Gloves - Straight Thumb

Made from premium leather to provide comfort and protection while on the job.

$15.54 - $149.00

Specifications

Color Natural
Glove Cuff Style Open
Grade Premium
Material Grain Cowhide Leather
Product Type Specs Drivers Gloves
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-200-390
View Documents
Wells Lamont™
Y0122S
Small
Dozen of 12 for $132.00
Case of 12 DZ for $1,584.00
Add to cart
 
17-200-391
View Documents
Wells Lamont™
Y0122M
Medium
Dozen of 12 for $132.00
Case of 12 DZ for $1,584.00
Add to cart
 
17-200-392
View Documents
Wells Lamont™
Y0122L
Large
Dozen of 12 for $15.54

Save 75% Reg : $62.17

Case of 12 DZ for $186.48

Save 88% Reg : $1,584.00

Add to cart
 
17-200-393
View Documents
Wells Lamont™
Y0122XL
X-Large
Dozen of 12 for $140.00
Case of 12 DZ for $1,680.00
Add to cart
 
17-200-394
View Documents
Wells Lamont™
Y0122XXL
2X-Large
Dozen of 12 for $149.00
Case of 12 DZ for $1,788.00
Add to cart
 
Description

Description

  • Abrasion resistance and insulation
  • Double shirred elastic wrist
  • Bound hem
  • Straight thumb
  • Gunn cut
Specifications

Specifications

Natural
Premium
Drivers Gloves
Gunn
Open
Grain Cowhide Leather
Y0122
Straight
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit