Learn More
American Scientific Grain of Wheat Led Bulb



Description
- Upgrade your outdated bulbs to the 21st century
- The wheat bulbs can also be used in control panels, high quality commercial and industrial controls
- Comes with a Teacher's Guide
- 0.003 A
- 3 V
- Pack of 100
Specifications
Specifications
| Description | Length: 8 in. |
| For Use With (Application) | Ideal for use in a variety of miniature equipment and circuit applications |
| Quantity | 100 Pk. |
| Weight (English) | 0.4 lb. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.