Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRAMD1A Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548654

Catalog No. PA548654


Add to cart

Description

Description

Predicted to react with rat based on sequence homology.

Specifications

Specifications

GRAMD1A
Polyclonal
PBS with 0.09% sodium azide
Q8VEF1
GRAM domain-containing protein 1A, GRAMD1A, KIAA1533
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
52857
Western Blot
Unconjugated
Gramd1a
Liquid
GRAMD1A
KLH conjugated synthetic peptide between 267-296 amino acids from the Central region of human GRAMD1A.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit