Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gramine, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
500g
Packaging:
Glass bottle
87-52-5
C11H14N2
174.25
MFCD00005629
OCDGBSUVYYVKQZ-UHFFFAOYSA-N
gramine,3-dimethylaminomethyl indole,donaxine,donaxin,gramin,1h-indole-3-methanamine, n,n-dimethyl,beta-dimethylaminomethylindole,1-1h-indol-3-yl-n,n-dimethylmethanamine,indol-3-ylmethyldimethylamine,1h-indol-3-ylmethyl dimethylamine
6890
CHEBI:28948
1-(1H-indol-3-yl)-N,N-dimethylmethanamine
CN(C)CC1=CNC2=CC=CC=C21
87-52-5
C11H14N2
174.25
MFCD00005629
OCDGBSUVYYVKQZ-UHFFFAOYSA-N
gramine,3-dimethylaminomethyl indole,donaxine,donaxin,gramin,1h-indole-3-methanamine, n,n-dimethyl,beta-dimethylaminomethylindole,1-1h-indol-3-yl-n,n-dimethylmethanamine,indol-3-ylmethyldimethylamine,1h-indol-3-ylmethyl dimethylamine
6890
CHEBI:28948
1-(1H-indol-3-yl)-N,N-dimethylmethanamine
CN(C)CC1=CNC2=CC=CC=C21
Sulfated Ash 0.5% max.
Quantity 100g
Loss on Drying 0.5% max.
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 131.0°C to 139.0°C
Color Beige
Flash Point 167°C
Assay Percent Range 99%
Merck Index 15, 4569
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,ether,toluene and chloroform, slightly soluble in cold acetone, practically insoluble in petr ether
Formula Weight 174.25
Physical Form Powder
Percent Purity 99%
Chemical Name or Material Gramine
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger