Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granisetron Hydrochloride, EP, 97-102%, Spectrum™

Click to view available options
Quantity:
100 g
Packaging:
Amber Glass Bottle

G2223, 107007-99-8, C18H25ClN4O

Spectrum™ Granisetron Hydrochloride, EP is used alone or with other medications to prevent nausea and vomiting caused by cancer drug treatment. All Spectrum Chemical EP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Loss on Drying 0.005
Sulfated Ash 0.001
Quantity 100 g
CAS 107007-99-8
CAS Max % 1
Molecular Formula C18H25ClN4O
InChI Key QYZRTBKYBJRGJB-UHFFFAOYNA-N
SMILES [H+].[Cl-].CN1N=C(C(=O)NC2CC3CCCC(C2)N3C)C2=CC=CC=C12
IUPAC Name hydrogen 1-methyl-N-{9-methyl-9-azabicyclo[3.3.1]nonan-3-yl}-1H-indazole-3-carboxamide chloride
Molecular Weight (g/mol) 348.88
Percent Purity 97 to 102%
Show More Show Less