Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granulin Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI3H6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270847JF549

Catalog No. N270847J549


Add to cart

Description

Description

Granulin Monoclonal antibody specifically detects Granulin in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Granulin
OTI3H6
Immunohistochemistry
Affinity Purified, Conjugated
GRN
Recombinant protein fragment corresponding to amino acids 327-593 of human Granulin (NP_002078) produced in E.coli.
Immunogen affinity purified
Cell Cycle and Replication
Primary
2896.0
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
acrogranin, GEP, GP88, granulin, granulin-epithelin, granulins, PC cell-derived growth factor, PCDGF, PEPI, PGRN, proepithelin, progranulin
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit