Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granzyme G Antibody (RM0247-12f21), Alexa Fluor™ 647, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212410AF647
Description
Granzyme G Monoclonal specifically detects Granzyme G in Mouse samples. It is validated for Western Blot.Specifications
Granzyme G | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GZMG | |
Mouse recombinant Granzyme G | |
0.1 mL | |
14944 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
RM0247-12f21 | |
Western Blot | |
Ctla7, Ctla-7 | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction