Learn More
Granzyme G Rat anti-Mouse, Alexa Fluor™ 350, Clone: RM0247-12f21, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212410AF350
Catalog No. NP212410A30
Add to cart
Description
Description
Granzyme G Monoclonal antibody specifically detects Granzyme G in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Granzyme G
RM0247-12f21
Western Blot
Ctla7, Ctla-7
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
GZMG
Mouse
0.1 mL
Primary
14944