Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRAP2 Antibody (OTI1G2), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271457PEATT594
Description
GRAP2 Monoclonal specifically detects GRAP2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
GRAP2 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
GRAP2 | |
Full length recombinant protein of human GRAP2 (NP_004801) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry | |
OTI1G2 | |
Flow Cytometry | |
Adapter protein GRID, GADSSH3-SH2-SH3 adapter Mona, GRB2L, GRB-2-like protein, GRB2-related adapter protein 2, GRB2-related adaptor protein 2, GRB2-related protein with insert domain, GRBLGGRAP-2, GRBX, grf-40, Grf40, Grf40 adapter protein, GRIDGRPL, Growth factor receptor-binding protein, growth factor receptor-bound protein 2-related adaptor protein 2, Hematopoietic cell-associated adapter protein GrpL, hematopoietic cell-associated adaptor protein GRPL, Mona, P38, Protein GADS, SH3-SH2-SH3 adaptor molecule | |
Mouse | |
Affinity Purified | |
Cancer | |
9402.0 | |
Store at 4°C in the dark. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction