Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Graphic Controls 4 in. Circular Charts for Dickson Recorders
Supplier: Graphic Controls 00166033
Specifications
Ink Writing Recorder Chart | |
114.3 mm | |
For Dickson recorder model: DTP, EV4, PR4, PR4-10, PW4, SK4 | |
Circular |
0 to 500, uniform along arc 50 to 450 by 100 and 100 to 400 by 100 | |
4.5 in. | |
2 | |
24 hrs. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction