Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Graphic Controls 4 in. Circular Charts for Dickson Recorders
Supplier: Graphic Controls 32012211
Specifications
Ink Writing Recorder Chart | |
114.3 mm | |
For Dickson recorder model: DTP, EV4, PR4, PR4-10, PW4, SK4, SL4, THP | |
Circular |
0 to 45, uniform along arc 5 to 35 by 10 and 10 to 40 by 10 | |
4.5 in. | |
2 | |
24 hrs. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction