Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Graphic Controls Recording Chart Paper: Hewlett-Packard™
Designed for clear and reliable data retrieval
Supplier: Graphic Controls 10645035
Description
This quality recording paper meets or exceeds manufacturers' specifications.
Specifications
Thermal Fanfold Recorder Chart Paper | |
Designed for clear and reliable data retrieval | |
184 ft. | |
8.42 in. |
184 ft. x 8.42 in. (56m x 213.86mm) | |
For recorder model: HP 3380A/S, 1080, 3385A, 5830A, 5840A, 5880 | |
56 m | |
213.86 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction