Learn More
Graphics Control Recording Charts, Circular
Manufacturer:  GRAPHIC CONTROLS ACQUISITION CORP W7703010
 View more versions of this product
Catalog No. 13-939-305
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
254mm
10 in.
1.0
-30° to -70°C
Ink Writing Recorder Chart
For chart recorder
Circular
7 Days