Alfa Aesar™ Graphite adhesive, alcohol based

Manufacturer: Alfa Aesar™ 4121315

Quantity 30g

 View more versions of this product

Catalog No. AA4121315

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00144065
    UN Number UN1133
    Chemical Name or Material Graphite adhesive, alcohol based, Service temp: 204°C

    Specifications

    Quantity 30g
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Flakes
    MDL Number MFCD00144065
    UN Number UN1133
    Health Hazard 1 Danger
    Particle Size 1 Micron
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H225-H318-H335-H336
    Highly flammable liquid and vapor.
    Causes serious eye damage.
    May cause respiratory irritation.
    May cause drowsiness or dizziness.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ADHESIVES
    Color Black
    Chemical Name or Material Graphite adhesive, alcohol based, Service temp: 204°C

    GHS H Statement
    H225-H318-H335-H336
    Highly flammable liquid and vapor.
    Causes serious eye damage.
    May cause respiratory irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit