Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graphite electrode, crater-drillpoint/undercut, 4.57mm dia, 38.10mm length, volume 0.040cc, 99.9995% (metals basis), Alfa Aesar™

Used as Homogeneous Catalysts. They are widely used in electric arc furnaces (EAF) and ladle furnaces (LF) for the melting of steel, non-ferrous metals and some non-metallic minerals.

Manufacturer:  Alfa Aesar 40788KN

Catalog No. AA40788KN


Order in Bulk

Request bulk or custom formats

Add to cart

Description

Description

Specifications

Specifications

Graphite electrode
100 pc.
AGKSP grade,Ultra "F" purity,ASTM S-14
Tube
Insoluble in water.
99.9995%
Safety and Handling

Safety and Handling

TSCA : U

Recommended Storage : Ambient temperatures

SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit