Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graphite felt, 99.9% (metal basis), Alfa Aesar™

Form: approx.9.2g/10 x 10cm

Manufacturer:  Alfa Aesar 14630RR

Quantity 10 x 10cm

 View more versions of this product

Catalog No. AA14630RR


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

Felt, 1.12cm (0.441 in.) thick
99.9% (Metal basis)
MFCD00144065
Graphite felt

Specifications

Dark grey to black solid in various forms
10 x 10cm
99.9% (Metal basis)
Graphite felt
Dark gray to black
Felt, 1.12cm (0.441 in.) thick
MFCD00144065

Safety and Handling

SDS & Safety Documents

EINECSNumber : 231-955-3

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit