Graphite Flake, Natural, -325 Mesh, 99.8% (Metal Basis), Alfa Aesar™

Manufacturer: Alfa Aesar 43209A1

Quantity 1kg

 View more versions of this product

Catalog No. AA43209A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7782-42-5
    Physical Form Flakes
    Assay Percent Range 99.8%, metal basis
    MDL Number MFCD00144065
    Chemical Name or Material Graphite, Natural

    Specifications

    Quantity 1kg
    DOT Information DOT Class: 4.1
    EINECS Number 231-955-3
    Physical Form Flakes
    Assay Percent Range 99.8%, metal basis
    MDL Number MFCD00144065
    CAS 7782-42-5
    Health Hazard 1 Flammable. Irritating to eyes and respiratory system.
    Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. This material and/or its container must be disposed of as hazardous waste.
    Chemical Name or Material Graphite, Natural

    In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. This material and/or its container must be disposed of as hazardous waste.

    Flammable. Irritating to eyes and respiratory system.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit