Alfa Aesar™ Graphite foil, 1mm (0.04 in.) thick, 97% (metals basis)

Manufacturer: Alfa Aesar™ 43083CJ

Quantity 30 x 100cm

 View more versions of this product

Catalog No. AA43083CJ

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00144065
    Chemical Name or Material Graphite foil, 1mm (0.04 in.) thick

    Specifications

    Quantity 30 x 100cm
    TSCA TSCA
    MDL Number MFCD00144065
    Physical Form Foil
    Chemical Name or Material Graphite foil, 1mm (0.04 in.) thick
    Assay 97%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit