Alfa Aesar™ Graphite Fusion Crucible
Manufactured from graphite materials that provide exceptional thermal uniformity and exactness to dimensional specifications.
Manufacturer: Alfa Aesar™ 14750UA
 View more versions of this product
Catalog No. AA14750UA
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Metallurgy
    Material Unpurified extruded graphite
    Quantity 15pc
    Shape Tapered
    Product Type Graphite fusion crucible
    Volume (English) 33.27cc
  • Description & Specifications
Related Products
Product Content Feedback