Learn More
Thermo Scientific Chemicals Graphite Gas Analysis Crucible; Ht (mm), 24.5; ID (mm), 10.0; OD (mm), 12.7; Tip Dia (mm), 6.80
Supplier:  Thermo Scientific Chemicals 039811KN
 View more versions of this product
Catalog No. AA39811KN
Order in Bulk
Request bulk or custom formats
Add to Cart
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10mm
For Gas analysis
24.5mm
6.8mm
Graphite
12.7mm
Graphite Crucible
Product Suggestions
Product Suggestions