Alfa Aesar™ Graphite ink for tantalum capacitors

Manufacturer: Alfa Aesar™ 42193A1

Quantity 1kg

 View more versions of this product

Catalog No. AA42193A1

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00144065
    Chemical Name or Material Graphite ink for tantalum capacitors

    Specifications

    Quantity 1kg
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H319-H335
    Causes serious eye irritation.
    May cause respiratory irritation.
    Physical Form Paste
    MDL Number MFCD00144065
    Health Hazard 1 Warning
    Color Black
    TSCA TSCA
    Odor Ammonia-like
    Chemical Name or Material Graphite ink for tantalum capacitors

    Description

    • A graphite ink for use as an undercoat for silver ink in the production of tantalum capacitors

    GHS H Statement
    H319-H335
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit