Learn More
Alfa Aesar™ Graphite plate, highly oriented pyrolytic graphite (HOPG), 10x10x(1.6min)mm (0.394x0.394x0.079in)
Manufacturer:  Alfa Aesar™ 43835KJ
Quantity 5 pieces
 View more versions of this product
Catalog No. AA43835KJ
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
MFCD00144065
Graphite plate, Highly oriented pyrolytic graphite (HOPG), 10x10x(1.6min)mm (0.394x0.394x0.079 in.)
Specifications
5 pieces
0.8 &Plusmn;0.2° Mosaic Angle
MFCD00144065
Graphite plate, Highly oriented pyrolytic graphite (HOPG), 10x10x(1.6min)mm (0.394x0.394x0.079 in.)
Safety and Handling
SDS & Safety Documents
TSCA : TSCA