Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graphite powder, crystalline, -300 mesh, 99%, Alfa Aesar

Synthetic from desulfurized petroleum coke

Manufacturer:  Alfa Aesar 1012914

Quantity 25g

 View more versions of this product

Catalog No. AA1012914


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7782-42-5
99%
MFCD00144065
Crystalline Powder
(CF)
Graphite

Specifications

Black powder
Black
7782-42-5
99%
MFCD00144065
Odorless
25g
Crystalline Powder
(CF)
Graphite

Safety and Handling

SDS & Safety Documents

H228-H319-H335: Flammable solid. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-955-3

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit