Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Graphite powder, natural, briquetting grade, -200 mesh, 99.9995% (metals basis)

Click to view available options
Quantity:
113g
227g
454g
7782-42-5
C
12.011
MFCD00144065
OKTJSMMVPCPJKN-UHFFFAOYSA-N
graphite,activated charcoal,norit,mineral,carbon-12,carbono,graphene,acticarbone,anthrasorb,carbosieve
5462310
CHEBI:27594
carbon
[C]
7782-42-5
C
12.011
MFCD00144065
OKTJSMMVPCPJKN-UHFFFAOYSA-N
graphite,activated charcoal,norit,mineral,carbon-12,carbono,graphene,acticarbone,anthrasorb,carbosieve
5462310
CHEBI:27594
carbon
[C]
Mesh Size −200 Mesh
Quantity 113g
Color Black
Odor Odorless
UN Number UN1325
Solubility Information Insoluble in water.
Physical Form Powder, UC-1 grade,Ultra "F" purity
Percent Purity 99.9995%
Grade UCP-1 grade, ultra 'F' purity
Chemical Name or Material Graphite powder, Natural
Assay (metals basis)
Show More Show Less

GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c

H228-H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 231-955-3

RTECSNumber : MD9659600

TSCA : Yes

Recommended Storage : Ambient temperatures