Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Graphite powder, synthetic, conducting grade, -325 mesh, 99.9995% (metals basis)
Click to view available options
Quantity:
28g
113g
227g
454g
Description
- Lubricants, pigments, anodes arc-lamp carbons, coating cathode ray tubes, moderator in nuclear piles
Specifications
Mesh Size | −325 Mesh |
Odor | Odorless |
Color | Black |
Quantity | 454g |
UN Number | UN1325 |
Solubility Information | Insoluble in water as well as other organic solvents. |
Physical Form | Powder, UC-2 grade,Ultra "F" purity |
Percent Purity | 99.9995% |
Grade | UCP-2 grade, ultra 'F' purity |
Chemical Name or Material | Graphite powder, Synthetic |
Show More |
Safety and Handling
GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H228-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 231-955-3
RTECSNumber : MD9659600
TSCA : Yes
Recommended Storage : Ambient temperatures