Graphite Rod, 1.27cm (0.5 in.) dia x 61cm (24 in.) long, 99% (Metals basis), Alfa Aesar™

Form: Approx. 130g/pc

Manufacturer: Alfa Aesar 10134KJ

Quantity 5 Pieces

 View more versions of this product

Catalog No. AA10134KJ

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Rod, 1.27cm (0.5 in.) dia. X 61cm (24 in.) long
    Assay Percent Range 99% (Metals basis)
    MDL Number MFCD00144065
    Chemical Name or Material Graphite

    Specifications

    Physical Form Rod, 1.27cm (0.5 in.) dia. X 61cm (24 in.) long
    Quantity 5 Pieces
    Color Dark gray to black
    MDL Number MFCD00144065
    Assay Percent Range 99% (Metals basis)
    Appearance Dark Gray to black rod
    Chemical Name or Material Graphite

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit