Alfa Aesar™ Graphite rod, 5cm (2.0 in.) dia. x 61cm (24 in.) long, 99% (metals basis)

Manufacturer: Alfa Aesar™ 00580KF

Quantity 1 piece

 View more versions of this product

Catalog No. AA00580KF

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    MDL Number MFCD00144065
    Chemical Name or Material Graphite rod, 5cm (2.0 in.) dia. x 61cm (24 in.) long

    Specifications

    Quantity 1 piece
    TSCA TSCA
    Physical Form Rod
    MDL Number MFCD00144065
    Weight ≈2.2kg/pc
    Chemical Name or Material Graphite rod, 5cm (2.0 in.) dia. x 61cm (24 in.) long
    Assay 99%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit