Learn More
Grasshoppers
SCI_ED
Brachystola sp.
Supplier:  Ranaco Corporation 126.1873.100
 View more versions of this product
Catalog No. S1875S100
Add to Cart
Description
Description
  • Lubber grasshoppers we use are all large, “Grade A” specimens
  • Each internally injected with a special formulation that ensures flexibility and preservation
  • Single-injected grasshoppers have their tracheal systems filled with colored latex
  • Size: 3 in. (7.6cm)
Specifications
Specifications
Grasshopper
3 in.
100 Pk.
Plain
7.6 cm
Pail
Product Suggestions
Product Suggestions