Learn More
Sklar InstrumentsSupplier Diversity Partner Sklar Black Graves Wide View Speculum, Sklar, 4 in. x 1 1/4 in. Stainless Steel EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 909057
Sklar Black Graves Wide View Speculum, Sklar, 4 in. x 1 1/4 in. Sklar Black Graves Wide View Speculum, Medium, With Loop To Hold and Direct Disposable Smoke Evacuation Tubing, Stainless Steel with Reduced Reflectivity Polymer Coating. The Sklar Black Graves Wide-View Vaginal Speculum is used for laser gynecological procedures. It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The Sklar Black Graves Vaginal Speculum is available with a permanent smoke evacuation tube, a loop for disposable smoke evacuation tubing, or no smoke evacuation. Sklar Black Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries.
Catalog No. 50-143-6917