Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Grb2/sem5/ASH Signaling Protein Ab-4, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.

Specifications

Antigen Grb2/sem5/ASH Signaling Protein Ab-4
Clone 2GB04; same as C1.4
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1457R1
View Documents
Thermo Scientific™
MS1457R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Grb2/sem5/ASH Signaling Protein Ab-4, Mouse Monoclonal Antibody.

Grb2 protein contains one SH2 domain flanked on both sides by an SH3 domain. Ab-1 binds to the junction between the amino terminal SH3 domain and the SH2 domain of Grb2. Reactivity with Grb2 deletion mutants suggests that its epitope maps between aa 54-62. In response to EGF, Grb2 binds, through its SH3 domains, to the guanine nucleotide exchange factor, Sos. The Grb2-Sos complex then binds the EGF-receptor. These interactions play a critical role in the activation of the p21ras signalling pathway.

Host Species: Mouse

Clone: 2GB04 (C1.4)

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Mouse, Rat and Xenopus. Others not known.

Epitope: aa 54-62

Immunogen: Purified recombinant human Grb2 protein

Molecular Weight: 24-25 kDa

Positive Control: HeLa cells, Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

General References:

1. Schlessinger J. Current Opinion in Genetics and Development, 1994 Feb, 4(1):25-30.

2. Boulikas T. Critical Reviews in Eukaryotic Gene Expression, 1995, 5(1):1-77.

3. Chardin P et al. Febs Letters, 1995 Aug 1, 369(1):47-51.

4. Downward J. Febs Letters, 1994 Jan 31, 338(2):113-7.

b>General References:

1. Schlessinger J. Current Opinion in Genetics and Development, 1994 Feb, 4(1):25-30.

2. Boulikas T. Critical Reviews in Eukaryotic Gene Expression, 1995, 5(1):1-77.

3. Chardin P et al. Febs Letters, 1995 Aug 1, 369(1):47-51.

4. Downward J. Febs Letters, 1994 Jan 31, 338(2):113-7.

Specifications

Specifications

Grb2/sem5/ASH Signaling Protein Ab-4
Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Primary
IgG2a, κ
2GB04; same as C1.4
Monoclonal
Murine
Signal Transduction
Purified recombinant human Grb2 protein
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit