Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GREB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550673
Description
The antibody detects endogenous levels of total GREB1 protein.
This gene is an estrogen-responsive gene that is an early response gene in the estrogen receptor-regulated pathway. It is thought to play an important role in hormone-responsive tissues and cancer. Three alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
GREB1 | |
Polyclonal | |
Unconjugated | |
GREB1 | |
5730583K22Rik; 9130004E13; AF180470; AU023194; gene regulated by estrogen in breast cancer; gene regulated by estrogen in breast cancer protein; gene regulated in breast cancer 1 protein; GREB1; growth regulating estrogen receptor binding 1; growth regulating estrogen receptor binding 1; protein GREB1; growth regulation by estrogen in breast cancer 1; id:ibd5087; id:ibd5087 protein; im:7140582; Kiaa0575; mKIAA0575; protein GREB1; protein GREB1-like; RGD1564091; si:ch211-175f11.3; zgc:158681 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9687 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q4ZG55 | |
GREB1 | |
Synthetic peptide corresponding to a region derived from internal residues of human growth regulation by estrogen in breast cancer 1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction