Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GREB1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310341100UL
Description
GREB1 Polyclonal specifically detects GREB1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GREB1 | |
Polyclonal | |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
gene regulated by estrogen in breast cancer protein, growth regulation by estrogen in breast cancer 1, KIAA0575Gene regulated in breast cancer 1 protein, protein GREB1 | |
The immunogen is a synthetic peptide directed towards the middle region of human GREB1 (NP_683701). Peptide sequence DNGHNALLGFSGNCVGCGKKGFCYFTEFSNHINLKLTTQPKKQKHLKYYL | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
9687 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction