Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Green 15 mil Unlined Chemical Resistant Nitrile Gloves

Excellent grip with wet and dry applications

$26.50

Specifications

Certifications/Compliance ISO 9000, ISO 9001:2000, 21 CFR 170-199, FDA
Color Green
Length (English) 13 in.
Length (Metric) 33 cm
Material Nitrile
View More Specs

Products 2
Catalog Number Size Price Quantity & Availability  
Catalog Number Size Price Quantity & Availability  
19-161-876
View Documents
Small
Dozen of 12 for $26.50
Case of 12 DZ for $318.00
Add to cart
 
19-161-879
View Documents
X-Large
Dozen of 12 for $26.50
Case of 12 DZ for $318.00
Add to cart
 
Description

Description

Acceptable for use in both food and pharmaceutical applications.

  • Embossed raised-diamond pattern on the palm and fingers
  • Size is embossed on each glove
  • Manufactured in an ISO 9002 certified facility
  • Color: green

Specifications

Specifications

ISO 9000, ISO 9001:2000, 21 CFR 170-199, FDA
13 in.
Nitrile
15 mil
Green
33 cm
Straight
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit