Learn More
Description
- Required to activate color marker functionality in Genesys software (version (1.X.X) and onward)
- For use in Gbox Chemi and Gbox Mini Systems
Specifications
Specifications
| Product Type | Visualization Filter |
| Color | Green |
| For Use With (Equipment) | Gbox Chemi and Gbox Mini Systems |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

