Learn More
Green Fluorescent Protein Rabbit anti-All, Unconjugated, Clone: 4I10, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB109725UL
 View more versions of this product
Catalog No. ZRB109725MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Green Fluorescent Protein Monoclonal antibody specifically detects Green Fluorescent Protein in All samples. It is validated for Immunocytochemistry, Immunofluorescence, Western Blotting.
Specifications
Specifications
Green Fluorescent Protein
4I10
Unconjugated
GFP
Rabbit
IgG
Protein A Purified
Secondary and Control Antibodies
Primary
All
Immunocytochemistry, Immunofluorescence, Western Blot
300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Purified
GFP
His-tagged full-length recombinant Green fluorescent protein (GFP) from Aequorea victoria (Jelly fish).
∼30 kDa observed; 26.89 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal