Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Green Fluorescent Protein Rabbit anti-All, Unconjugated, Clone: 4I10, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen Green Fluorescent Protein
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB109725MI
View Documents
MilliporeSigma
ZRB109725UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB10974XMI
View Documents
MilliporeSigma
ZRB10974X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Green Fluorescent Protein Monoclonal antibody specifically detects Green Fluorescent Protein in All samples. It is validated for Immunocytochemistry, Immunofluorescence, Western Blotting.
Specifications

Specifications

Green Fluorescent Protein
Unconjugated
Rabbit
GFP
His-tagged full-length recombinant Green fluorescent protein (GFP) from Aequorea victoria (Jelly fish).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunofluorescence, Western Blot
Purified
All
GFP
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit