Learn More
Eisco™ Green Slate Metamorphic Rock Specimen Products for Science Education
Green slate is a very lightly metamorphosed and well-foliated specimen. It can be distinguished from it's parent rock (Shale) by the difference in pitch when struck.
$.50 - $6.00
Specifications
Grade K-16
Type Metamorphic Rock
Specimen Size 2-3 cm
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
S27593 Products for Science Education
View Documents
Eisco™
ESNG0086
1/Ea. Each for $0.50
Add to cart
S27615 Products for Science Education
View Documents
Eisco™
ESNG0086PK12
12/Pk. Pack of 12 for $6.00
Add to cart
Description
Description
  • Correlates well to Next Generation Science Standards Evidence
  • Perfect for lessons on metamorphic rocks and alteration
  • Ideal for identification & classification exercises
Specifications
Specifications
K-16
Metamorphic Rock
2-3 cm