Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Green Tea Extract, 4:1, Powder, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
84650-60-2
84650-60-2

G1281, 84650-60-2

Spectrum™ Green Tea Extract, 4:1, Powder
Packaging Amber Glass Bottle
Quantity 100 g
Loss on Drying 0.09
CAS Max % 1