Learn More
Qorpak™ Green Thermoset Cap with F217 and PTFE Liner
Click to view available options
Neck Finish:
13-425
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
8-425
Packaging:
Bagged
Bulk
Description
- The color green easily identifies PTFE liner system
- F-217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature
- Provides odor and impact resilience for a tight seal with a low moisture transmission rate
- F-217 has a low density foam core between two solid layers of LDPE (low density polyethylene)
- Available in smooth and ribbed style
Specifications
Closure Liner | F217, PTFE |
Closure Type | Smooth |
Closure Material | Thermoset |
Material | Thermoset |
No. per Case | 5300 |
Packaging | Bulk |
Closure Color | Green |
Neck Finish | 22-400 |