Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte GREEN Water Monitoring Kit
An inquiry-based approach to watershed education.
$278.00
Specifications
For Use With (Application) | Environmental education |
---|---|
Grade | 4+ |
Includes | Biochemical oxygen demand, coliform bacteria, dissolved oxygen, nitrate, pH, phosphate, turbidity, temperature, diagrammed instructions, color chart, test factor flashcard, all necessary apparatus and nonhazardous testabs™ reagents |
Includes: Set of nine test factor modules, each including: diagrammed instructions, color chart, test factor flashcard, all necessary apparatus and nonhazardous TesTabs reagents to perform 100 tests (44 tests for coliform bacteria)
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S65402
![]() |
LaMotte
5848 |
Green Water Monitoring Kit |
Each for $278.00
|
|
Description
- Great tool for beginning a school-based, community-involved water monitoring program
- Learn to design and implement a river study through the use of watershed maps
- Compile and analyze results, share the results with your community and learn to take action through a variety of productive avenues
Order Info
Grades 4+
Specifications
Environmental education | |
4+ | |
Biochemical oxygen demand, coliform bacteria, dissolved oxygen, nitrate, pH, phosphate, turbidity, temperature, diagrammed instructions, color chart, test factor flashcard, all necessary apparatus and nonhazardous testabs™ reagents |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title