Greiner Bio-One Hematology K3 EDTA Evacuated Tubes
Manufacturer: Greiner Bio-One 454012
 View more versions of this product
Catalog No. 22-040-187
  Check Availability
Add to Cart
    Specifications
    Material Polyethylene terephthalate
    Additive Potassium solution
    Color Black ring, Pink cap
    Diameter (Metric) Outer 13x75mm
    Capacity (Metric) 4mL
    Closure Type Safety Pull Cap
  • Description & Specifications
Related Products
Product Content Feedback