Learn More
Greiner Bio-One Multi-Drawing Blood Collection Needles
For routine blood collection
Manufacturer:  Greiner Bio-One 450048
 View more versions of this product
Catalog No. 22-030-650
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
CE, OSHA
Routine Blood collection
Polypropylene
1.5 in.
Synthetic Rubber
Stainless Steel
Yes
Yes
18g
Pink protective cap, safety valve at one end
3.81cm
4 to 25°C
Vacuette™ Multi Sample Blood Collection Needle