Learn More
Greiner Bio-One Plasma Tubes with Lithium Heparin and Gel
For determinations in clinical chemistry and immunochemistry
Manufacturer:  Greiner Bio-One 456087
 View more versions of this product
Catalog No. 22-040-028
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
  • Interior tube wall is coated with lithium heparin, an anticoagulant that binds antithrombins to block the coagulation cascade producing a plasma sample
  • Contain a barrier gel in the tube that moves upward to form a stable barrier separating plasma from cells during centrifugation; plasma may be aspirated directly from the tube eliminating the need for transfer
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • Tubes have an 18-month shelf life from the date of manufacture
  • Nonscrew Safety Pull Caps can be removed with a gentle pull
Description & Specifications
Specifications
5mL
Lithium Heparin
13 x 100mm
Safety Pull Cap
Yellow ring, Green cap
13 x 100mm